Learn More
Cole-Parmer Styrofoam R-8000 storage box for 100 (1.5 to 2.0 mL) microcentrifuge tubes
Supplier:  Cole-Parmer 4435020
Encompass
This STYROFOAM storage box holds 1.5- and 2.0-mL microcentrifuge tubes. The box has alphanumerically indexed tube cavities that hold 100 tubes, organized in five rows of 20. Snug-fitting lid has a finger notch for easy opening. The box is made of recyclable expanded polystyrene. Shpg wt 0.8 lb (0.4 kg).
Catalog No. NC9618614