Learn More
Thermo Scientific SHISEIDO GUARD CARTRIDGE MG
Catalog No. NC9972926 Shop All Thermo Scientific Products
Encompass
Guard Cartridge, Capcell PAK MG, C18, 4.0 x 20 mm, 3um (Pkg/2)
Catalog No. NC9972926 Supplier Thermo Scientific Supplier No. 8812307
Guard Cartridge, Capcell PAK MG, C18, 4.0 x 20 mm, 3um (Pkg/2)