Learn More
Aviva Systems Biology Corp Shkbp1 Antibody - N-terminal region (ARP35400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Shkbp1. It was validated on Western Blot. . Alternate names: B930062H15Rik, Sb1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-461