Learn More
Med Vet International Adjustable Mobile X-Ray Shield, Royal Blue, J1725, Each
Catalog No. 5000495932
Encompass_Preferred
The standard of care for taking X-Rays is moving towards hands-free for clinic safety. This mobile X-Ray stand is easily portable while shielding staff. Features: • Adjustable from 24" to 48" (61cm to 122cm) • Shield width 24" (61cm) • 0.5mm Pb equivalency • Chrome frame • Stable caster base
Catalog No. 50-004-95932 Supplier Med Vet International Supplier No. JOR1725
The standard of care for taking X-Rays is moving towards hands-free for clinic safety. This mobile X-Ray stand is easily portable while shielding staff. Features: • Adjustable from 24" to 48" (61cm to 122cm) • Shield width 24" (61cm) • 0.5mm Pb equivalency • Chrome frame • Stable caster base