Learn More
Electron Microscopy Sciences EMS Shield Mounting Medium with (DAPI) & Pr 100 ML
SDP
Catalog No. 501927427
Encompass_Preferred
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Phycoerythrin (RP-E), Phycocyanin (PC), and Allophycocyanin (APC). Fluorescence is retained during prolonged storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC.
This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI.
Catalog No. 50-192-7427 Supplier Electron Microscopy Sciences Supplier No. 1798931
EMS Shield with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears. This unique formula prevents rapid photobleaching of FITC, Texas Red, AMCA, Cy2, Cy3, Cy5, Alexa fluoro 488, Alexa fluoro 594, Green fluorescent protein (GFP), tetramethyly rhodamine, Redox. Phycoerythrin (RP-E), Phycocyanin (PC), and Allophycocyanin (APC). Fluorescence is retained during prolonged storage at 4 degree C in the dark. This medium does not contain phenylenediamine, which destroys immunofluorescence of Cy dyes, RP-E, PC and APC.
This mounting medium is fortified with DAPI which is a counter-stain for DNA. This product is to be used in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360nm and emits at 460nm, producing a blue fluorescence. RNA is also stained with DAPI.