Learn More
Caron Products And Services Additional Stainless Steel Perforated Shelf Kit. Includes 2 shelf tracks.
Catalog No. NC2293874
Encompass_Preferred
Additional Stainless Steel Perforated Shelf Kit. Includes 2 shelf tracks. For 50 ft3 and 75 ft3 chambers.
This item is not returnable. View return policy
Catalog No. NC2293874 Supplier Caron Products And Services Supplier No. SHLF325
Additional Stainless Steel Perforated Shelf Kit. Includes 2 shelf tracks. For 50 ft3 and 75 ft3 chambers.