Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SHMT-IN-1 | 1508286-18-7 | 99.86% | 10 MG
SDP

Catalog No. 5000339438
Encompass_Preferred

SHMT-IN-1 (compound (±)-46) is a potent inhibitor of plasmodial serine hydroxymethyltransferase (SHMT) and exhibits antitumor activity.

  • Potent inhibitor of plasmodial serine hydroxymethyltransferase (SHMT).
  • Exhibits antitumor activity.
  • Molecular weight: 375.25.
  • Formula: C18H16Cl2N4O.
  • Appearance: Solid.
  • Color: White to off-white.
  • Target: SHMT.
  • Pathway: Metabolic enzyme/protease.
  • Solubility in DMSO: 250 mg/mL (666.22 mM).

Catalog No. 50-003-39438 Supplier Medchemexpress LLC Supplier No. HY12965110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SHMT-IN-1 (compound (±)-46) is a potent inhibitor of plasmodial serine hydroxymethyltransferase (SHMT) and exhibits antitumor activity.

  • Potent inhibitor of plasmodial serine hydroxymethyltransferase (SHMT).
  • Exhibits antitumor activity.
  • Molecular weight: 375.25.
  • Formula: C18H16Cl2N4O.
  • Appearance: Solid.
  • Color: White to off-white.
  • Target: SHMT.
  • Pathway: Metabolic enzyme/protease.
  • Solubility in DMSO: 250 mg/mL (666.22 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.