Learn More
TARGETMOL CHEMICALS INC SHMT-IN-1 5MG
SDP
Catalog No. 502581404
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SHMT-IN-1 is a potent plasmodial serine hydroxymethyltransferase (SHMT) inhibitor with antitumor activity. purity: 99%
Catalog No. 50-258-1404 Supplier TARGETMOL CHEMICALS INC Supplier No. T12902
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SHMT-IN-1 is a potent plasmodial serine hydroxymethyltransferase (SHMT) inhibitor with antitumor activity. purity: 99%