Learn More
Aviva Systems Biology Corp SHMT1 Antibody - N-terminal region (ARP49040_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49040_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHMT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SHMT1. Alternate names: CSHMT, MGC15229, MGC24556, SHMTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-62962