Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci SHMT1 Recombinant Protein
SDP

Catalog No. 16593117
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-593-117 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-593-117 Supplier ProSci Supplier No. 916870050
Only null left
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Serine Hydroxymethyltransferase Cytosolic (SHMT1) is a member of the SHMT family. SHMT1 is a cytoplasmic protein and exists as a homotetramer. SHMT1 catalyzes the reversible conversion of serine and tetrahydrofolate to glycine and 5,10-methylene tetrahydrofolate. This reaction provides one carbon unit for the synthesis of methionine, thymidylate, and purines in the cytoplasm. A reduction in SHMT1 levels would result in less glycine that could affect the nervous system by acting as an agonist to the NMDA receptor and this could be a mechanism behind Smith-Magenis syndrome.

Specifications

Accession Number P34896
Gene ID (Entrez) 6470
Molecular Weight (g/mol) 53.9 kDa
Quantity 0.05 mg
Source Human Cells
Common Name serine hydroxymethyltransferase 1
Gene Symbol SHMT1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Met3-Phe483
Content And Storage Lyophilized protein should be stored at -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4°C to 7°C for 2-7 days. Aliquots of reconstituted samples are stable (at -20°C for 3 months.)
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Serine Hydroxymethyltransferase Cytosolic, SHMT, Glycine Hydroxymethyltransferase, Serine Methylase, SHMT1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.