Learn More
Aviva Systems Biology Corp SHMT2 Antibody - C-terminal region (ARP46129_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46129_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SHMT2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human SHMT2. Alternate names: GLYA, SHMTApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59240