Learn More
Cayman Chemical ShnzhIsIde methyl ester 10mg
SDP
Catalog No. 502950127
Encompass_Preferred
An iridoid glycoside with anti-inflammatory and antinociceptive activities; a GLP-1R agonist; inhibits the release of MPO and elastase from fMLP-activated isolated rat neutrophils at 10 µM; inhibits LPS-induced increases in the release of IL-8 and LTB4 from isolated rat neutrophils at 10 µM; reduces L5/L6 spinal nerve ligation-induced mechanical allodynia in rats (ED50 = 40.4 UG/animal)
Catalog No. 50-295-0127 Supplier Cayman Chemical Supplier No. 3404710 MG
An iridoid glycoside with anti-inflammatory and antinociceptive activities; a GLP-1R agonist; inhibits the release of MPO and elastase from fMLP-activated isolated rat neutrophils at 10 µM; inhibits LPS-induced increases in the release of IL-8 and LTB4 from isolated rat neutrophils at 10 µM; reduces L5/L6 spinal nerve ligation-induced mechanical allodynia in rats (ED50 = 40.4 UG/animal)