Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Shock-Absorbing Lanyard 3 ft 3

Catalog No. 502795124
Encompass_Preferred

Shock-Absorbing Lanyard 3 ft 310 lb.

Catalog No. 50-279-5124 Supplier Grainger Supplier No. 19Z839
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Shock-Absorbing Lanyard 3 ft 310 lb.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.