Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ SHOCK-STOP™ Lockout Hasps

Catalog No. 19104140 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Silver
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-140 Silver
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-104-140 Supplier Brady™ Supplier No. 87693
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-visibility metal hasp with 6 lock holes

Specifications

Product Type Scissor-action Hasp
Color Silver
Material Steel
Certifications/Compliance OSHA 1910.147
Depth (English) 0.178 in.
Dimensions (D x W x H) 0.178 x 3.468 x 5.456 in.
Height (English) 5.456 in.
Quantity 5 Pk.
Width (English) 3.468 in.
Width (Metric) 99.06 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.