Learn More
MSC Milwaukee Tool SHOCKWAVE Impact Duty Magnetic Bit Holder, PH2 Phillips Insert Bits, 2-3/8 in. Length, 5 Pack
Catalog No. 5000510566
Encompass
The SHOCKWAVE™ Impact Duty 2.36 in. magnetic bit tip holder with Phillips #2 insert bits is designed for use with impact drivers and drill drivers. This accessory features a durable construction and is engineered to provide an unyielding bit engagement and maximum contact for demanding applications.
  • Absorbs peak torque and prevents breaking with optimized shank geometry
  • Protects fit over the life of the bit with enhanced wear resistance
  • Reduces stripping and wobble via a precise fit custom machined tip
  • Engineered for extended life with proprietary steel and custom heat treatment
  • Includes powerful magnet for secure grip
  • Features laser etched size for easy identification
Catalog No. 50-005-10566 Supplier MSC Supplier No. 90693714
The SHOCKWAVE™ Impact Duty 2.36 in. magnetic bit tip holder with Phillips #2 insert bits is designed for use with impact drivers and drill drivers. This accessory features a durable construction and is engineered to provide an unyielding bit engagement and maximum contact for demanding applications.
  • Absorbs peak torque and prevents breaking with optimized shank geometry
  • Protects fit over the life of the bit with enhanced wear resistance
  • Reduces stripping and wobble via a precise fit custom machined tip
  • Engineered for extended life with proprietary steel and custom heat treatment
  • Includes powerful magnet for secure grip
  • Features laser etched size for easy identification