Learn More
Waters Corp Waters Column, Reversed Phase, Polymer, 6 μm, 6 mm x 150 mm
Catalog No. 50785343 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is a polymeric reversed-phase column designed for high-performance liquid chromatography. It is stable within a broad pH range of 2 to 12 and primarily supports legacy methods, composed of porous polymeric particles.
  • Polymeric reverse phase columns.
  • Designed for applications requiring a broad pH range of 2 to 12.
  • Sorbent functions similarly to traditional polymer-based materials, suitable for more hydrophobic than hydrophilic samples.
  • Polymethacrylate packing performs effectively with mobile phases containing relatively high concentrations of water.
  • Consistently delivers excellent quality and desired results for liquid chromatography.
  • Stable in a pH range of 2 to 12.
  • Features a base polymer of Polystyrene.
Catalog No. 50-785-343 Supplier Waters Corp Supplier No. WAT034220
This column is a polymeric reversed-phase column designed for high-performance liquid chromatography. It is stable within a broad pH range of 2 to 12 and primarily supports legacy methods, composed of porous polymeric particles.
  • Polymeric reverse phase columns.
  • Designed for applications requiring a broad pH range of 2 to 12.
  • Sorbent functions similarly to traditional polymer-based materials, suitable for more hydrophobic than hydrophilic samples.
  • Polymethacrylate packing performs effectively with mobile phases containing relatively high concentrations of water.
  • Consistently delivers excellent quality and desired results for liquid chromatography.
  • Stable in a pH range of 2 to 12.
  • Features a base polymer of Polystyrene.