Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Shodex IC 5u YK-421 20A

Catalog No. 502412815 Shop All Phenomenex Products
Encompass

Guard Column 10 x 4.6 mm

Catalog No. 50-241-2815 Supplier Phenomenex Inc Supplier No. YKG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guard Column 10 x 4.6 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.