Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, SEC/GPC, 5 μm, 8 mm x 300 mm

Catalog No. 50785326 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This product is an ultrahigh efficiency GPC column primarily supporting legacy methods. It serves as an Organic SEC (GPC) Column for general analysis. Ideal for investigating particles sized 5 μm, it complies with USP L21 criteria. The column provides high speeds and resolution, separating organics and oligomers by molecular size using rigid polystyrene gels and an organic solvent. It also measures the molecular weight distribution of polymers, aiding in polymer component separation.

  • Separation mode: SEC/GPC
  • pH range: 2 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Particle shape: Spherical
  • Packing solvent: Chloroform
  • Ideal for investigating particles sized 5 μm
  • Separates organics and oligomers by molecular size
  • Measures molecular weight distribution of polymers

Catalog No. 50-785-326 Supplier Waters Corp Supplier No. WAT034111
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an ultrahigh efficiency GPC column primarily supporting legacy methods. It serves as an Organic SEC (GPC) Column for general analysis. Ideal for investigating particles sized 5 μm, it complies with USP L21 criteria. The column provides high speeds and resolution, separating organics and oligomers by molecular size using rigid polystyrene gels and an organic solvent. It also measures the molecular weight distribution of polymers, aiding in polymer component separation.

  • Separation mode: SEC/GPC
  • pH range: 2 to 12
  • Maximum pressure: 6000 psi (415 bar)
  • Particle shape: Spherical
  • Packing solvent: Chloroform
  • Ideal for investigating particles sized 5 μm
  • Separates organics and oligomers by molecular size
  • Measures molecular weight distribution of polymers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.