Learn More
Waters Corp Shodex Guard Column, 7 μm, 6 mm, 50 mm
Catalog No. 50785359 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard column primarily supports legacy methods and is designed to be used with the KC-811 column.
  • Primarily supports legacy methods.
  • Guard column for KC-811 column.
  • Features a polymer particle substrate.
  • Operates effectively within a pH range of 2 to 12.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Designed with spherical particle shape.
  • Equipped with a Parker-style endfitting.
  • Suitable for HPLC systems.
Catalog No. 50-785-359 Supplier Waters Corp Supplier No. WAT035501
This guard column primarily supports legacy methods and is designed to be used with the KC-811 column.
  • Primarily supports legacy methods.
  • Guard column for KC-811 column.
  • Features a polymer particle substrate.
  • Operates effectively within a pH range of 2 to 12.
  • Withstands a maximum pressure of 6000 psi (415 Bar).
  • Designed with spherical particle shape.
  • Equipped with a Parker-style endfitting.
  • Suitable for HPLC systems.