Learn More
Waters Corp Shodex Column, 6 μm, 8 mm × 300 mm
Catalog No. 50785314 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column primarily supports legacy methods and is an ultrahigh efficiency GPC column.
  • Primarily supports legacy methods.
  • Ultrahigh efficiency GPC column.
  • Accommodates a wide range of applications from low to high molecular weight compounds.
  • Allows polymer scientists to evaluate polymer physical properties by analyzing the molecular weight distribution of the polymer.
  • Provides information about specific physical qualities based on the predicted molecular weight distribution.
  • Can be used to separate a polymer from smaller oligomers, monomer forms, and other formulation ingredients.
  • Satisfies industry standards for great performance and efficiency.
Catalog No. 50-785-314 Supplier Waters Corp Supplier No. WAT030699
This column primarily supports legacy methods and is an ultrahigh efficiency GPC column.
  • Primarily supports legacy methods.
  • Ultrahigh efficiency GPC column.
  • Accommodates a wide range of applications from low to high molecular weight compounds.
  • Allows polymer scientists to evaluate polymer physical properties by analyzing the molecular weight distribution of the polymer.
  • Provides information about specific physical qualities based on the predicted molecular weight distribution.
  • Can be used to separate a polymer from smaller oligomers, monomer forms, and other formulation ingredients.
  • Satisfies industry standards for great performance and efficiency.