Learn More
Waters Corp Shodex Column, 6 μm, 8 mm x 300 mm
Catalog No. 50785313 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Shodex KF-802 Column is designed for high-speed and high-resolution analysis, primarily supporting legacy methods. This ultrahigh efficiency GPC column is perfectly suited for separating organics and oligomers by molecular size, utilizing an organic solvent as eluent and hard polystyrene gels as packing material. It performs molecular weight distribution measurements of polymers and supports a wide range of applications from low to high molecular weight compounds, enabling polymer scientists to characterize polymer physical properties. As an Organic SEC (GPC) Column, it can be used for general analysis with THF and fulfills USP L21 requirements.
  • High-speed and high-resolution analysis
  • Separation of organics and oligomers by molecular size
  • Measurement of the molecular weight distribution of polymers
  • Meets high performance and efficiency benchmarks
  • Fulfills USP L21 requirements
Catalog No. 50-785-313 Supplier Waters Corp Supplier No. WAT030698
The Shodex KF-802 Column is designed for high-speed and high-resolution analysis, primarily supporting legacy methods. This ultrahigh efficiency GPC column is perfectly suited for separating organics and oligomers by molecular size, utilizing an organic solvent as eluent and hard polystyrene gels as packing material. It performs molecular weight distribution measurements of polymers and supports a wide range of applications from low to high molecular weight compounds, enabling polymer scientists to characterize polymer physical properties. As an Organic SEC (GPC) Column, it can be used for general analysis with THF and fulfills USP L21 requirements.
  • High-speed and high-resolution analysis
  • Separation of organics and oligomers by molecular size
  • Measurement of the molecular weight distribution of polymers
  • Meets high performance and efficiency benchmarks
  • Fulfills USP L21 requirements