Learn More
Waters Corp Shodex Column, 6 μm, 8 mm x 300 mm
Catalog No. 50785318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Shodex KF-803 is an Organic SEC (GPC) Column designed for high-speed, high-resolution analysis and legacy methods. It uses hard polystyrene gels with narrow particle size distribution and organic solvents as eluent. This column separates organics and oligomers by molecular size and measures polymer molecular weight distribution for various compounds, meeting high manufacturing and performance standards.
  • High-speed and high-resolution analysis, supports legacy methods.
  • Uses organic solvents as eluent with hard polystyrene gels (narrow particle size).
  • Separates organics and oligomers by molecular size, measures polymer molecular weight distribution.
  • Suitable for general analysis (THF) and diverse low/high molecular weight compounds.
  • Meets high manufacturing, performance, and efficiency standards.
  • Fulfills USP L21 requirements.
Catalog No. 50-785-318 Supplier Waters Corp Supplier No. WAT034100
The Shodex KF-803 is an Organic SEC (GPC) Column designed for high-speed, high-resolution analysis and legacy methods. It uses hard polystyrene gels with narrow particle size distribution and organic solvents as eluent. This column separates organics and oligomers by molecular size and measures polymer molecular weight distribution for various compounds, meeting high manufacturing and performance standards.
  • High-speed and high-resolution analysis, supports legacy methods.
  • Uses organic solvents as eluent with hard polystyrene gels (narrow particle size).
  • Separates organics and oligomers by molecular size, measures polymer molecular weight distribution.
  • Suitable for general analysis (THF) and diverse low/high molecular weight compounds.
  • Meets high manufacturing, performance, and efficiency standards.
  • Fulfills USP L21 requirements.