Learn More
Waters Corp Shodex Column, SEC/GPC, 7 μm, 8 mm x 300 mm
Supplier:  Waters Corp WAT034101
Encompass_Preferred
This ultrahigh efficiency GPC column supports legacy methods, separating organics and oligomers by molecular size. It uses an organic solvent as eluent and hard polystyrene gels as packing, making it suitable for diverse applications with varying molecular compound weights.
  • Supports legacy methods and offers ultrahigh efficiency GPC.
  • Separates organics and oligomers by molecular size using organic solvents and polystyrene gels.
  • Suitable for a wide range of molecular compound weights.
  • Meets high performance, efficiency, and USP L21 requirements.
  • Optimized for 7 μm particle size analysis; delivers high-speed, high-resolution results.
  • Enables polymer molecular weight distribution measurement.
  • Facilitates polymer separation from smaller oligomers, monomers, and other additives.
  • Effective across a wide pH range of 2 to 12.
Catalog No. 50-785-319