Learn More
Waters Corp Waters Column, 7 μm, 8 mm x 300 mm
Catalog No. 50785355 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column primarily supports legacy methods and is part of the polymeric columns for sugar and carbohydrate analysis. It is suitable for various applications where molecular compound weights range from low to high, offering separation based on differences in analytes’ coordination complex. The column’s stationary phase is modified with a metal sulfonate complex ion and works in combination with size exclusion or HILIC modes.
  • Supports legacy methods.
  • Designed for sugar and carbohydrate analysis.
  • Suitable for applications with a wide range of molecular compound weights.
  • Separates based on differences in analytes’ coordination complex.
  • Stationary phase modified with metal sulfonate complex ion.
  • Works with size exclusion or HILIC modes.
Catalog No. 50-785-355 Supplier Waters Corp Supplier No. WAT034277
This column primarily supports legacy methods and is part of the polymeric columns for sugar and carbohydrate analysis. It is suitable for various applications where molecular compound weights range from low to high, offering separation based on differences in analytes’ coordination complex. The column’s stationary phase is modified with a metal sulfonate complex ion and works in combination with size exclusion or HILIC modes.
  • Supports legacy methods.
  • Designed for sugar and carbohydrate analysis.
  • Suitable for applications with a wide range of molecular compound weights.
  • Separates based on differences in analytes’ coordination complex.
  • Stationary phase modified with metal sulfonate complex ion.
  • Works with size exclusion or HILIC modes.