Learn More
Waters Corp Shodex Column, 7 μm, 8 mm x 300 mm, Polymer
Catalog No. 50785356 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Shodex KS-804 Column is designed to primarily support legacy methods for sugar and carbohydrate analysis, utilizing polymeric columns. It is an HPLC column with a spherical polymer particle substrate.
  • Suitable for protein applications.
  • Characterized by its robust pH range of 2 to 12.
  • Specifications include an 8 mm inner diameter.
  • 300 mm length.
  • Particle size of 7 μm.
Catalog No. 50-785-356 Supplier Waters Corp Supplier No. WAT034279
The Shodex KS-804 Column is designed to primarily support legacy methods for sugar and carbohydrate analysis, utilizing polymeric columns. It is an HPLC column with a spherical polymer particle substrate.
  • Suitable for protein applications.
  • Characterized by its robust pH range of 2 to 12.
  • Specifications include an 8 mm inner diameter.
  • 300 mm length.
  • Particle size of 7 μm.