Learn More
Waters Corp Waters Shodex KW-802.5 HPLC Column, 5 μm, 8 mm × 300 mm, 400 Å
Supplier: Waters Corp WAT035943
The Shodex KW-802.5 column is a high-performance gel filtration chromatography (GFC) column for separating proteins, enzymes, and polysaccharides. It features a 400Å pore size and excellent chemical stability (pH 2-8), suitable for protein and peptide analysis. Its packing material is porous, spherical silica gel with hydrophilic hydroxyl groups, ensuring minimal swell/shrinkage and compatibility with various buffer solutions or polar organic solvents.
- High-performance gel filtration chromatography (GFC)
- Separates proteins, enzymes, and polysaccharides
- Features 5 μm spherical silica particles
- Dimensions: 8 mm ID x 300 mm length
- Pore size: 400 Å
- Stable across pH range of 2-8
- Exclusion limits: 60,000 (pullulan) and 150,000 (proteins)
- Diol (OH) chemistry
- Meets USP L20, L33, and L59
- Parker-style endfittings
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.