Learn More
Waters Corp Shodex Column, 5 μm, 8 mm, 300 mm, 1000 Å
Catalog No. 50785381 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-performance Gel Filtration column provides high-speed and high-resolution analysis, ideal for separating proteins, enzymes, and polysaccharides. It uses spherical silica gel packing with hydrophilic hydroxy groups for chemical stability (pH 3-7.5) and minimal swell. Suitable for 1000Å pore size analysis, and meets USP L20, L33, L59 requirements.
  • High-speed and high-resolution analysis.
  • Designed for high-performance gel filtration chromatography (GFC).
  • Optimized for protein, enzyme, and polysaccharide separation.
  • Silica-based stationary phases for excellent protein and peptide separation.
  • Uses porous, spherical silica gel packing with hydrophilic hydroxy groups.
  • Minimal swell and shrinkage, allowing various buffer solutions or polar organic solvents.
  • Chemical stability for pH 3-7.5.
  • Meets USP L20, L33, and L59 requirements.
Catalog No. 50-785-381 Supplier Waters Corp Supplier No. WAT035946
Add to Cart
Add to Cart
This high-performance Gel Filtration column provides high-speed and high-resolution analysis, ideal for separating proteins, enzymes, and polysaccharides. It uses spherical silica gel packing with hydrophilic hydroxy groups for chemical stability (pH 3-7.5) and minimal swell. Suitable for 1000Å pore size analysis, and meets USP L20, L33, L59 requirements.
  • High-speed and high-resolution analysis.
  • Designed for high-performance gel filtration chromatography (GFC).
  • Optimized for protein, enzyme, and polysaccharide separation.
  • Silica-based stationary phases for excellent protein and peptide separation.
  • Uses porous, spherical silica gel packing with hydrophilic hydroxy groups.
  • Minimal swell and shrinkage, allowing various buffer solutions or polar organic solvents.
  • Chemical stability for pH 3-7.5.
  • Meets USP L20, L33, and L59 requirements.