Learn More
Waters Corp Waters Corporation Shodex Pullulan Kit, GFC Standards, 8 for Molecular Weight Determination
Catalog No. 50785339 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit offers a unique collection of 8 GFC standards for molecular weight determination in aqueous environments. It supports GPC, GFC, and SEC separation modes with RI/ELSD and UV detection methods, making it suitable for gel filtration applications.
  • Unique collection of 8 GFC standards for molecular weight determination.
  • Used for aqueous molecular weight testing, gel filtration, GPC, GFC, and SEC separation modes.
  • Supports RI/ELSD and UV detection methods.
  • Sample solutions are stable, preventing agglomeration, crystallization, or electrical charge.
  • Pullulan is not adsorbed to column packing materials, suitable for Size Exclusion Chromatography calibration.
  • Contains 0.2 g each of 8 approximate molecular weight standards from 5000 to 800000.
  • Solid formulation with a 36-month shelf life at 4°C; one week after reconstitution.
Catalog No. 50-785-339 Supplier Waters Corp Supplier No. WAT034207
This kit offers a unique collection of 8 GFC standards for molecular weight determination in aqueous environments. It supports GPC, GFC, and SEC separation modes with RI/ELSD and UV detection methods, making it suitable for gel filtration applications.
  • Unique collection of 8 GFC standards for molecular weight determination.
  • Used for aqueous molecular weight testing, gel filtration, GPC, GFC, and SEC separation modes.
  • Supports RI/ELSD and UV detection methods.
  • Sample solutions are stable, preventing agglomeration, crystallization, or electrical charge.
  • Pullulan is not adsorbed to column packing materials, suitable for Size Exclusion Chromatography calibration.
  • Contains 0.2 g each of 8 approximate molecular weight standards from 5000 to 800000.
  • Solid formulation with a 36-month shelf life at 4°C; one week after reconstitution.