Learn More
Waters Corp Shodex Column, 6 μm, 8 mm, 300 mm
Catalog No. 50785350 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polymeric column supports legacy methods for sugar analysis, fulfilling USP L19 and L22 requirements for sugar separation.
  • Packed with rigid styrene/DVB-based strong cation-exchange resin.
  • Employs size-exclusion, partition, and ligand exchange chromatography for separation.
  • Separates saccharides via complex configuration differences between hydroxyl groups and metal ions.
  • Utilizes Ca2+, Pb2+, and Na+ counter ions.
  • Requires only water for neutral sugar analysis.
  • For ionic substances, a salt (Ca(NO3)2 or CaSO4) with pH 3 to 7 should be added.
  • Ca-EDTA (10 to 50 ppm) prevents performance deterioration with heavy metal samples.
  • Suitable for food, biochemical, and natural product applications.
  • Delivers high-performance saccharide separation.
Catalog No. 50-785-350 Supplier Waters Corp Supplier No. WAT034238
This polymeric column supports legacy methods for sugar analysis, fulfilling USP L19 and L22 requirements for sugar separation.
  • Packed with rigid styrene/DVB-based strong cation-exchange resin.
  • Employs size-exclusion, partition, and ligand exchange chromatography for separation.
  • Separates saccharides via complex configuration differences between hydroxyl groups and metal ions.
  • Utilizes Ca2+, Pb2+, and Na+ counter ions.
  • Requires only water for neutral sugar analysis.
  • For ionic substances, a salt (Ca(NO3)2 or CaSO4) with pH 3 to 7 should be added.
  • Ca-EDTA (10 to 50 ppm) prevents performance deterioration with heavy metal samples.
  • Suitable for food, biochemical, and natural product applications.
  • Delivers high-performance saccharide separation.