Learn More
Waters Corp Waters Column, 7 μm, 8 mm, 300 mm, 1/pk
Supplier:  Waters Corp WAT036954
Encompass_Preferred
This Shodex high performance polymeric column is designed for sugar analysis, supporting legacy methods. It enables analysis of saccharides and sugar alcohols in a single run with a single eluent, often requiring no organic solvent. The column uses a rigid styrene/DVB-based strong cation exchange resin, employing size-exclusion chromatography, partition chromatography, and ligand exchange for sugar separation.
  • Supports legacy methods.
  • High performance polymeric column for sugar analysis.
  • Analyzes saccharides and sugar alcohols in a single run with a single eluent.
  • Utilizes size-exclusion, partition, and ligand exchange chromatography.
  • Requires only water for neutral sugar analysis.
  • Compatible with sample analysis equipment.
Catalog No. 50-785-417