Learn More
Protexer Corporation Shoe Cover Refill - Non-skid, Non-woven, Polypropylene, 40g-600pairs/case w/Rubberband
Catalog No. NC1487735
Encompass_Preferred
Constructed of durable spunbond polypropylene material with non-skid, non-conductive soles. Skid-resistant pattern improves friction and traction. Strong elastic band around ankle allows for a universal fit. One Size fits most up to men’s size 15. Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
This item is not returnable. View return policy
Catalog No. NC1487735 Supplier Protexer Corporation Supplier No. KBNS40SF600
Constructed of durable spunbond polypropylene material with non-skid, non-conductive soles. Skid-resistant pattern improves friction and traction. Strong elastic band around ankle allows for a universal fit. One Size fits most up to men’s size 15. Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.