Learn More
Protexer Corporation Shoe Cover Refill - Polyethylene - Case - 825 pairs
Catalog No. NC1229351
Encompass_Preferred
Constructed of Chlorinated polyethylene (CPE), this waterproof bootie is an ideal low-cost solution for light-duty application. Strong elastic band around ankle allows for a universal fit. One size fits most up to men’s size 15. Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.
This item is not returnable. View return policy
Catalog No. NC1229351 Supplier Protexer Corporation Supplier No. KBPE825
Constructed of Chlorinated polyethylene (CPE), this waterproof bootie is an ideal low-cost solution for light-duty application. Strong elastic band around ankle allows for a universal fit. One size fits most up to men’s size 15. Approved for Class 100 & Class 5 Cleanroom use per Federal Standard 209E and ISO STD. 14664-1 Class 5. For use with Large and Small Kinetic BootieButler Dispensers.