Learn More
Med Vet International Versapro Disposable Polypropylene Shoe Covers, Blue - Bagged, 1000/Case
Catalog No. 502829362
Encompass_Preferred
Economical and disposable Made of polypropylene non woven fabric1000 blue shoe covers in each case The cover fits shoes up to size 10Keep sanitary-sensitive areas free and clear of foreign substances Remove and trap unwanted dirt, dust and debris from shoes Help improve indoor air quality Polyethylene Shoe Covers are made of a low density polyethylene film making them liquid impervious and lint-free. These shoe covers are an economical alternative when a low particulate material is needed to protect against splash.
Catalog No. 50-282-9362 Supplier Med Vet International Supplier No. PESC
Economical and disposable Made of polypropylene non woven fabric1000 blue shoe covers in each case The cover fits shoes up to size 10Keep sanitary-sensitive areas free and clear of foreign substances Remove and trap unwanted dirt, dust and debris from shoes Help improve indoor air quality Polyethylene Shoe Covers are made of a low density polyethylene film making them liquid impervious and lint-free. These shoe covers are an economical alternative when a low particulate material is needed to protect against splash.