Learn More
Med Vet International Shoe Covers, Xl, 100/Box
Catalog No. 502830297
Encompass_Preferred
Latex-free, non-skid, multi-layer shoe covers. Fluid-resistant and strong. The tread on the bottom provides excellent traction. Generously sized for full-coverage protection. Sport size fits almost any athletic shoe. Regular fits up to men's size 12; XL fits up to men's size 15
Catalog No. 50-283-0297 Supplier Med Vet International Supplier No. SHOECOVERXL
Latex-free, non-skid, multi-layer shoe covers. Fluid-resistant and strong. The tread on the bottom provides excellent traction. Generously sized for full-coverage protection. Sport size fits almost any athletic shoe. Regular fits up to men's size 12; XL fits up to men's size 15