Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Shorr Staining Solution

Catalog No. M1092750500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1092750500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1092750500 Supplier MilliporeSigma™ Supplier No. 1.09275.0500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the diagnosis of hormonal dysfunction. MilliporeSigma™ Shorr Staining Solution is a ready-to-use solution that makes it possible to evaluate cytological target structures in human-gynecological specimen materials, for example cervical smears of vag

TRUSTED_SUSTAINABILITY

Specifications

Color Nuclei: brownish red, Eosinophilic cytoplasm: red, Cyanophilic cytoplasm: blue-green
Description Ready to use Shorr staining solution, sufficient for 1000 stainings
For Use With (Application) For hormonal cyto-diagnosis of vaginal epithelium
Packaging Plastic bottle
Quantity 500 mL
Storage Requirements Store at 15°–25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.