Learn More
Azzota Corp 10mm Standard Glass Cuvette - 1.5Ml Half Height Of Regular Cuvettes Pathlength - 10mm Inside Width - 10mm Volume - 1.5Ml Dimensions - 22.5X12.5X12.5mm Cover And Lid Optical Glass (340Nm-2500Nm)
Catalog No. 507532879
Encompass_Preferred
10mm Standard Glass Cuvette - 1.5ml half height of regular cuvettes Pathlength - 10mm Inside width - 10mm Volume - 1.5ml Dimensions - 22.5x12.5x12.5mm Cover and lid Optical Glass (340nm-2500nm)
Catalog No. 50-753-2879 Supplier Azzota Corp Supplier No. G104S1
10mm Standard Glass Cuvette - 1.5ml half height of regular cuvettes Pathlength - 10mm Inside width - 10mm Volume - 1.5ml Dimensions - 22.5x12.5x12.5mm Cover and lid Optical Glass (340nm-2500nm)