Learn More
Vernier™ Short Calculator Link Cable
SDPSCI_ED
Catalog No. S16267ND Shop All Vernier Software Products
Change view
Click to view available options
Length (English):
6 in.
Catalog No. Length (English)
S16267ND 6 in.
1 options
Catalog No. S16267ND Supplier Vernier™ Supplier No. TISLC
Includes
Short Calculator Link Cable
This 6 in. cable is used to connect a LabPro or CBL 2 to the I/O port of TI-73, TI-83, TI-83 Plus, TI-86, or TI-89 calculator. The cable has the small I/O plug on both ends. This cable is included with a CBL 2 or LabPro interface.
  • The cable has the small I/O plug on both ends.
  • This cable is included with a CBL 2 or LabPro interface.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Equipment) CBL 2 and LabPro
Length (English) 6 in.
Length (Metric) 15.24 cm
For Use With (Application) Educational use only