Learn More
MSC Proto Combination Wrench, 1/4 in, 12-Point, Full Polish, 1 ea
Catalog No. 501920137
Encompass
This Proto combination wrench is engineered for efficient and durable performance in diverse fastening tasks. It features a 12-point box end that enhances speed and access, making it compatible with 4, 6, and 12-point fasteners, and has a full polish finish for corrosion resistance.
  • A 12-point box end allows for fine indexing and improved access
  • Fits 4, 6, and 12-point fasteners
  • Self-aligns with the fastener for ease of use
  • Countersunk box end for easy application
  • A 15° angled offset helps clear obstructions
  • Anti-slip Design (ASD) open end
  • Box ends incorporate the TorquePlus™ system
  • Made from alloy steel for durability
Catalog No. 50-192-0137 Supplier MSC Supplier No. 39942867
This Proto combination wrench is engineered for efficient and durable performance in diverse fastening tasks. It features a 12-point box end that enhances speed and access, making it compatible with 4, 6, and 12-point fasteners, and has a full polish finish for corrosion resistance.
  • A 12-point box end allows for fine indexing and improved access
  • Fits 4, 6, and 12-point fasteners
  • Self-aligns with the fastener for ease of use
  • Countersunk box end for easy application
  • A 15° angled offset helps clear obstructions
  • Anti-slip Design (ASD) open end
  • Box ends incorporate the TorquePlus™ system
  • Made from alloy steel for durability