Learn More
Perkin Elmer US LLC Short Focal Length Mirror Kit - GPOB Accessory
Catalog No. 502113103
Encompass_Preferred
The General Purpose Optical Bench (GPOB) accessory is compatible with the LAMBDA 650/850/850+/950/1050/1050+ spectrometers. The GPOB comes with either a PbS detector (PELA1003) or an InGaAs detector (L6020328). Also available are a variety of optical kits that expand GPOB analysis. Features and benefits of this unique accessory: The GPOB specializes in the measurement of difficult or irregular samples. It enables custom optical measurement configurations using a magnetized, ruled optical bench with a movable 60 mm integrating sphere. Typical configurations include transmittance measurements of large laser crystals or telephoto lenses, variable-angle transmittance and reflectance studies of large glass samples, and beam splitter characterization. It can accommodate samples up to 400 mm in pathlength. Wavelength range is 1902500 nm.
Catalog No. 50-211-3103 Supplier Perkin Elmer US LLC Supplier No. PELA1013
The General Purpose Optical Bench (GPOB) accessory is compatible with the LAMBDA 650/850/850+/950/1050/1050+ spectrometers. The GPOB comes with either a PbS detector (PELA1003) or an InGaAs detector (L6020328). Also available are a variety of optical kits that expand GPOB analysis. Features and benefits of this unique accessory: The GPOB specializes in the measurement of difficult or irregular samples. It enables custom optical measurement configurations using a magnetized, ruled optical bench with a movable 60 mm integrating sphere. Typical configurations include transmittance measurements of large laser crystals or telephoto lenses, variable-angle transmittance and reflectance studies of large glass samples, and beam splitter characterization. It can accommodate samples up to 400 mm in pathlength. Wavelength range is 1902500 nm.