Learn More
Poltex PETG Short Glove Box Holder
SDP
Catalog No. 22000360
Change view
Click to view available options
Capacity:
1 Box
2 Boxes
3 Boxes
4 Boxes
Mounting:
Keyhole Slots
Magnetic Mount
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Mounting
22-000-360 1 Box Keyhole Slots
22-000-364 1 Box Magnetic Mount
22-000-361 2 Boxes Keyhole Slots
22-000-365 2 Boxes Magnetic Mount
22-000-362 3 Boxes Keyhole Slots
22-000-366 3 Boxes Magnetic Mount
22-000-363 4 Boxes Keyhole Slots
22-000-367 4 Boxes Magnetic Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 22-000-360 Supplier Poltex Supplier No. 1GBSHORTW
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
Holders are constructed from extremely durable, clear PETG with a lift-off face and short stature to allow for mounting in compact spaces. Front face hooks on with a secure hold when gloves are dispensing, but easily lifts off to replace empty boxes.
  • Constructed from extremely durable, clear PETG plastic
  • Lift-off face and short stature allows for mounting in compact spaces
  • Front face hooks on with a secure hold, but easily lifts off to replace empty glove boxes
  • Mounting hardware is not included for keyhole mount model
Specifications
Product Type Glove Box Holder
Color Clear
Dimensions (D x W x H) Interior 3.7 x 5.5 x 7 in.
Holds Disposable Glove Boxes
Material PETG
Mounting Keyhole Slots
No. of Compartments 1
Product Style Top Loading
Capacity 1 Box