Learn More
Perkin Elmer US LLC Short Graphite/Vespel Ferrules - 0.0625 in x 0.4 mm I.D., Pkg. 10
Catalog No. 502111825
Encompass_Preferred
Short ferrules for use with the Clarus 590/690 Capillary Injector at temperatures up to 350 °C.
Catalog No. 50-211-1825 Supplier Perkin Elmer US LLC Supplier No. 09200685
Short ferrules for use with the Clarus 590/690 Capillary Injector at temperatures up to 350 °C.