Learn More
MSC Vikan Hand Brush, Ultra Safe Technology, Stiff Bristle, Short Handle, Orange
Catalog No. 502838247
Encompass
This hand brush features durable polypropylene construction with stiff polyester bristles, designed to minimize contamination and bristle loss through its angled filament security units. It is ideal for cleaning conveyor belts, machinery, and food prep surfaces in high-risk areas. The brush includes an ergonomic handle with a hole for convenient hanging storage and is color-coded to reduce the risk of cross-contamination. This product meets the safety requirements for plastic materials intended to come into contact with food as set out by the European Framework Regulation (EU 10/2011).
  • Features angled filament security units to minimize contamination and bristle loss.
  • Ergonomic handle includes a hole for hanging storage.
  • Color-coded design helps reduce cross-contamination.
  • Certified for EU 10/2011 on food contact, ensuring safety for food-related applications.
Catalog No. 50-283-8247 Supplier MSC Supplier No. 54807144
This hand brush features durable polypropylene construction with stiff polyester bristles, designed to minimize contamination and bristle loss through its angled filament security units. It is ideal for cleaning conveyor belts, machinery, and food prep surfaces in high-risk areas. The brush includes an ergonomic handle with a hole for convenient hanging storage and is color-coded to reduce the risk of cross-contamination. This product meets the safety requirements for plastic materials intended to come into contact with food as set out by the European Framework Regulation (EU 10/2011).
  • Features angled filament security units to minimize contamination and bristle loss.
  • Ergonomic handle includes a hole for hanging storage.
  • Color-coded design helps reduce cross-contamination.
  • Certified for EU 10/2011 on food contact, ensuring safety for food-related applications.