Learn More
MSC Vikan Washing Brush, Short Handle, Soft/split, 10.6 in, Green
Catalog No. 502837573
Encompass
This Washing Brush features an ergonomic short handle and soft/split bristles, enhancing cleaning efficacy by retaining water and chemicals. It is ideal for fragile surfaces like glass and Perspex, and designed for critical hygiene and food safety environments.
  • Soft/split bristles prevent surface damage and hold more water and cleaning chemicals.
  • Ergonomic design with finger grips enhances comfort and allows multiple holding positions.
  • Durable construction for long-lasting daily performance.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.
  • Suitable for cleaning fragile surfaces like glass and Perspex.
  • Ideal for food manufacturing, food retail, restaurants, and food service.
Catalog No. 50-283-7573 Supplier MSC Supplier No. 47175997
This Washing Brush features an ergonomic short handle and soft/split bristles, enhancing cleaning efficacy by retaining water and chemicals. It is ideal for fragile surfaces like glass and Perspex, and designed for critical hygiene and food safety environments.
  • Soft/split bristles prevent surface damage and hold more water and cleaning chemicals.
  • Ergonomic design with finger grips enhances comfort and allows multiple holding positions.
  • Durable construction for long-lasting daily performance.
  • Easy to clean and maintain for hygiene control.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Drop-shaped hanging hole prevents liquid pooling and eases storage.
  • Suitable for cleaning fragile surfaces like glass and Perspex.
  • Ideal for food manufacturing, food retail, restaurants, and food service.