Learn More
Cole-Parmer Analytik Jena Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 115 VAC/60 Hz
Catalog No. NC9475680 Shop All Cole-Parmer Products
Encompass_Preferred
This compact, short and long wave lamp comes with two, four watt tubes (254 and 365 nm). Each tube produces a distinct wavelength without mixing of primary UV ranges. Order optional lamp stand 09817-00 separately.
Catalog No. NC9475680 Supplier Cole-Parmer Supplier No. 0981720
This compact, short and long wave lamp comes with two, four watt tubes (254 and 365 nm). Each tube produces a distinct wavelength without mixing of primary UV ranges. Order optional lamp stand 09817-00 separately.