Learn More
MSC Vikan Mixing Paddle with Holes, Polypropylene, 40 inch, Blue
Catalog No. 502838152
Encompass
This two-in-one tool is useful for both mixing and scraping tasks in food processing, such as soup, confectionery, dough, and sauce manufacturing. Its one-piece construction and long handle design make it easy to use and reduce user fatigue, offering a lightweight plastic alternative to metal mixing paddle scrapers. It is also easy to clean and maintain for hygiene control.
  • Designed for food manufacturing, food retail, restaurants, and food service.
  • Versatile two-in-one tool for mixing and scraping.
  • Long handle for easier and more ergonomic reach into tanks and vats.
  • Durable construction for long-lasting performance.
  • Easy to clean and maintain.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Complies with FDA Regulation CFR 21.
  • Complies with California Proposition 65.
  • Halal and Kosher compliant.
Catalog No. 50-283-8152 Supplier MSC Supplier No. 47171236
This two-in-one tool is useful for both mixing and scraping tasks in food processing, such as soup, confectionery, dough, and sauce manufacturing. Its one-piece construction and long handle design make it easy to use and reduce user fatigue, offering a lightweight plastic alternative to metal mixing paddle scrapers. It is also easy to clean and maintain for hygiene control.
  • Designed for food manufacturing, food retail, restaurants, and food service.
  • Versatile two-in-one tool for mixing and scraping.
  • Long handle for easier and more ergonomic reach into tanks and vats.
  • Durable construction for long-lasting performance.
  • Easy to clean and maintain.
  • Color-coded for hygienic zoning plans and 5S lean programs.
  • Complies with FDA Regulation CFR 21.
  • Complies with California Proposition 65.
  • Halal and Kosher compliant.