Learn More
Ace Glass, Inc. Short path still, 500mL, complete with distillation flask, gas inlet tube with adapter, 24/40 still body, thermometer, cow receiver and (3)250mL receiving flasks
SDP
Catalog No. 50973254
Encompass_Preferred
24/40 jointed short path still. Extension tube from still body allows positioning of drop directly over receiver flask. Gas inlet tube is positioned in the distillation flask with an ACE 5028 adapter and secured with a #7 Ace-Thred. Thermometer joint is 10/30. Complete system consists of 500mL distillation flask, still body, cow receiver, gas inlet tube, threaded adapter, 3 250mL receiver flasks and a thermometer.
Catalog No. 50-973-254 Supplier Ace Glass, Inc. Supplier No. 655448
24/40 jointed short path still. Extension tube from still body allows positioning of drop directly over receiver flask. Gas inlet tube is positioned in the distillation flask with an ACE 5028 adapter and secured with a #7 Ace-Thred. Thermometer joint is 10/30. Complete system consists of 500mL distillation flask, still body, cow receiver, gas inlet tube, threaded adapter, 3 250mL receiver flasks and a thermometer.