Learn More
Analytical Sales & Services Short PEEK Flangeless Fitting with Ferrule, 1/4-28, for 1/16" Tubing
Catalog No. 5000805845
Encompass
Short PEEK Flangeless Fitting with Ferrule\n• Thread: 1/4-28\n• For 1/16″ OD tubing\n\nQty/Pkg: 10PK\n\n \n
Catalog No. 50-008-05845 Supplier Analytical Sales & Services Supplier No. 97235X
Short PEEK Flangeless Fitting with Ferrule\n• Thread: 1/4-28\n• For 1/16″ OD tubing\n\nQty/Pkg: 10PK\n\n \n