Learn More
Gel Company Short Plate, for Mini-PROTEAN 3 & Tetra Cell, Package of 5
Catalog No. NC0256330
Encompass_Preferred
Short plate (7 x 10 cm), for Mini-PROTEAN 3 & Tetra Cell, pkg of 5.
Catalog No. NC0256330 Supplier Gel Company Supplier No. GBU3S5
Short plate (7 x 10 cm), for Mini-PROTEAN 3 & Tetra Cell, pkg of 5.