Learn More
Chemglass Life Sciences Reaction Vessel Lid, Short, 6-Nk, 250mL, Equivalent to Mettler Multimax Part # 51161188
SDP
Catalog No. 501532043
Encompass_Preferred
Reaction Vessel Lid, Short, 6-Nk, 250mL, Compatible with Mettler Systems. Compatible with Mettler Systems: 14/20 CN, (1) 14/20 SN, (2) 19/22 SN and (2) GL-14 SN's; 98mm OD Flange with Notch; Equivalent to Mettler Part # 51161188
Catalog No. 50-153-2043 Supplier Chemglass Life Sciences Supplier No. CGMR250L1
Reaction Vessel Lid, Short, 6-Nk, 250mL, Compatible with Mettler Systems. Compatible with Mettler Systems: 14/20 CN, (1) 14/20 SN, (2) 19/22 SN and (2) GL-14 SN's; 98mm OD Flange with Notch; Equivalent to Mettler Part # 51161188