Learn More
Medicus Health Short Test Tube Adapters for Centrifuge, 10 PK
SDP
Catalog No. NC2152057
Encompass
∙ Short tube adapters fit in bottom of tube shield make short tubes easier to remove from the centrifuge∙ Great for 4mL and 7mL Vacutainer Tubes∙ Available as a package of 10∙ Each adapter measures: 1.5"H x .5"W
This item is not returnable. View return policy
Catalog No. NC2152057 Supplier Medicus Health Supplier No. 5261M3
∙ Short tube adapters fit in bottom of tube shield make short tubes easier to remove from the centrifuge∙ Great for 4mL and 7mL Vacutainer Tubes∙ Available as a package of 10∙ Each adapter measures: 1.5"H x .5"W